Butyric acid, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W222100-10KG-K W222100-1KG-K W222100-25KG-K
SIALW222100-10KG-KEA 209 EUR
SIALW222100-10KG-K SIALW222100-1KG-K SIALW222100-25KG-K
Butyric acid, Sigma-Aldrich®
Butyric acid
Formula: C₄H₈O₂
Boiling Pt: 163.5 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0.958 g/cm³ (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: 8,III
Merck Index: 13,01593