Lauric acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Dodecanoic acid, Dodecylic acid
M-Clarity™ quality level = MQ300
W261408-10KG-K W261408-1KG-K W261408-25KG-K W261408-5KG-K
SIALW261408-10KG-KEA 230 EUR
SIALW261408-10KG-K SIALW261408-1KG-K SIALW261408-25KG-K SIALW261408-5KG-K
Lauric acid, Sigma-Aldrich®
Lauric acid
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 296
Melting Pt: 42…45 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400