Acetic acid 96% 96
Supplier: Roth Carl
Danger
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
X895.1 X895.2 X895.3
ROTHX895.1EA 29.4 EUR
ROTHX895.1 ROTHX895.2 ROTHX895.3
Acetic acid 96% 96
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM