Heptanoic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Heptylic acid, Enanthic acid
M-Clarity™ quality level = MQ300
W334812-100G-K W334812-1KG-K
SIALW334812-100G-KEA 85 EUR
SIALW334812-100G-K SIALW334812-1KG-K
Heptanoic acid, Sigma-Aldrich®
Heptanoic acid
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004426
CAS Number: 111-14-8
EINECS: 203-838-7
UN: 3265
ADR: 8,III
Merck Index: 13,04680