Cerotic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

H0388-100MG H0388-25MG
SIALH0388-100MGEA 257 EUR
SIALH0388-100MG SIALH0388-25MG
Cerotic acid, Sigma-Aldrich®
Cerotic acid
Formula: C₂₆H₅₂O₂
Boiling Pt: 419 °C (1013 hPa)
Melting Pt: 76…77 °C
Density: 0.8198 g/cm³ (100 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002811
CAS Number: 506-46-7
EINECS: 208-040-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR