Caproic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Hexanoic acid
M-Clarity™ quality level = MQ200
153745-100G 153745-2.5G 153745-500G
SIAL153745-100GEA 42.4 EUR
SIAL153745-100G SIAL153745-2.5G SIAL153745-500G
Caproic acid, Sigma-Aldrich®
Caproic acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –4.8 °C
Density: 0.9276 g/cm³ (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00004421
CAS Number: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: 8,III
Merck Index: 13,01765