Caproic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Hexanoic acid
M-Clarity™ quality level = MQ300
W255904-1KG-K W255904-25KG-K W255904-5KG-K
SIALW255904-1KG-KEA 79 EUR
SIALW255904-1KG-K SIALW255904-25KG-K SIALW255904-5KG-K
Caproic acid, Sigma-Aldrich®
Caproic acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –4.8 °C
Density: 0.9276 g/cm³ (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00004421
CAS Number: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: 8,III
Merck Index: 13,01765