Pelargonic acid, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Nonanoic acid, Pelargonic Acid

M-Clarity™ quality level = MQ100

N29902-1L N29902-500ML
SIALN29902-1LEA 145 EUR
SIALN29902-1L SIALN29902-500ML
Pelargonic acid, Sigma-Aldrich®
Pelargonic acid
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 12 °C
Density: 0.9055 g/cm³ (20 °C)
Flash Pt: 133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004433
CAS Number: 112-05-0
EINECS: 203-931-2
UN: 3265
ADR: 8,III
Merck Index: 12,07198

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR