Pelargonic acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Nonanoic acid, Pelargonic Acid
M-Clarity™ quality level = MQ100
N5502-100G
N5502-25G
SIALN5502-100GEA
80.3
EUR
SIALN5502-100G
SIALN5502-25G
Pelargonic acid, Sigma-Aldrich®
Pelargonic acid
Formula:
C₉H₁₈O₂ MW: 158.24 g/mol Boiling Pt: 253 °C (1013 hPa) Melting Pt: 12 °C Density: 0.9055 g/cm³ (20 °C) Flash Pt: 133 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004433 CAS Number: 112-05-0 EINECS: 203-931-2 UN: 3265 ADR: 8,III Merck Index: 12,07198 |