Pelargonic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Nonanoic acid, Pelargonic Acid
M-Clarity™ quality level = MQ100
W278408-10KG-K W278408-1KG-K W278408-20KG-K W278408-5KG-K
SIALW278408-10KG-KEA 345 EUR
SIALW278408-10KG-K SIALW278408-1KG-K SIALW278408-20KG-K SIALW278408-5KG-K
Pelargonic acid, Sigma-Aldrich®
Pelargonic acid
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 12 °C
Density: 0.9055 g/cm³ (20 °C)
Flash Pt: 133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004433
CAS Number: 112-05-0
EINECS: 203-931-2
UN: 3265
ADR: 8,III
Merck Index: 12,07198