Stearic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W303518-1KG-K W303518-8KG-K
SIALW303518-1KG-KEA 76.4 EUR
SIALW303518-1KG-K SIALW303518-8KG-K
Stearic acid, Sigma-Aldrich®
Stearic acid
Formula: C₁₈H₃₆O₂
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 67 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM