Caprylic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid
M-Clarity™ quality level = MQ200
O3907-1L O3907-2L O3907-500ML
SIALO3907-1LEA 54.3 EUR
SIALO3907-1L SIALO3907-2L SIALO3907-500ML
Caprylic acid, Sigma-Aldrich®
Caprylic acid
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.9066 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771