Caprylic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid
M-Clarity™ quality level = MQ300
W279900-10KG-K W279900-20KG-K W279900-2KG-K
SIALW279900-10KG-KEA 338 EUR
SIALW279900-10KG-K SIALW279900-20KG-K SIALW279900-2KG-K
Caprylic acid, Sigma-Aldrich®
Caprylic acid
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.9066 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771