Valeric acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Pentanoic acid
M-Clarity™ quality level = MQ300
W310107-10KG-K W310107-1KG-K W310107-25KG-K W310107-5KG-K
SIALW310107-10KG-KEA 316 EUR
SIALW310107-10KG-K SIALW310107-1KG-K SIALW310107-25KG-K SIALW310107-5KG-K
Valeric acid, Sigma-Aldrich®
Valeric acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 186 °C (1013 hPa)
Melting Pt: –32 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00004413
CAS Number: 109-52-4
EINECS: 203-677-2
UN: 3265
ADR: 8,III
Merck Index: 12,10042