Lignoceric acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tetracosanoic acid
M-Clarity™ quality level = MQ200
L6641-100MG L6641-1G L6641-10G L6641-5G
SIALL6641-100MGEA 45.3 EUR
SIALL6641-100MG SIALL6641-1G SIALL6641-10G SIALL6641-5G
Lignoceric acid, Sigma-Aldrich®
Lignoceric acid
Formula: C₂₄H₄₈O₂
MW: 368.64 g/mol
Boiling Pt: 406
Melting Pt: 87
Density: 0.8207 g/cm³ (100 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002810
CAS Number: 557-59-5
EINECS: 209-180-7
Merck Index: 13,05509