Myristic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Tetradecylic acid, Tetradecanoic acid
M-Clarity™ quality level = MQ200
M3128-100G
M3128-10G
SIALM3128-100GEA
116
EUR
SIALM3128-100G
SIALM3128-10G
Myristic acid, Sigma-Aldrich®
Myristic acid
Formula:
C₁₄H₂₈O₂ Boiling Pt: 319 °C (1013 hPa) Melting Pt: 51…54 °C Density: 0.8622 g/cm³ (20 °C) Flash Pt: >165 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002744 CAS Number: 544-63-8 EINECS: 208-875-2 Merck Index: 12,06416 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...