Myristic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tetradecylic acid, Tetradecanoic acid
M-Clarity™ quality level = MQ100
W276405-10KG-K W276405-1KG-K W276405-25KG-K
SIALW276405-10KG-KEA 391 EUR
SIALW276405-10KG-K SIALW276405-1KG-K SIALW276405-25KG-K
Myristic acid, Sigma-Aldrich®
Myristic acid
Formula: C₁₄H₂₈O₂
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 51…54 °C
Density: 0.8622 g/cm³ (20 °C)
Flash Pt: >165 °C
Storage Temperature: Freezer
MDL Number: MFCD00002744
CAS Number: 544-63-8
EINECS: 208-875-2
Merck Index: 12,06416