Myristic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Tetradecylic acid, Tetradecanoic acid
M-Clarity™ quality level = MQ300
W276413-10KG-K
W276413-1KG-K
W276413-25KG-K
W276413-SAMPLE
SIALW276413-10KG-KEA
379
EUR
SIALW276413-10KG-K
SIALW276413-1KG-K
SIALW276413-25KG-K
SIALW276413-SAMPLE
Myristic acid, Sigma-Aldrich®
Myristic acid
Formula:
C₁₄H₂₈O₂ Boiling Pt: 319 °C (1013 hPa) Melting Pt: 51…54 °C Density: 0.8622 g/cm³ (20 °C) Flash Pt: >165 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002744 CAS Number: 544-63-8 EINECS: 208-875-2 Merck Index: 12,06416 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...