Cesium acetate ≥99.998% (metals basis)
Supplier: Thermo Scientific
Synonyms: Acetic acid cesium salt
44434.14 44434.22
44434.14EA 125 EUR
44434.14 44434.22
Cesium acetate ≥99.998% (metals basis)
Cesium acetate
Purity: 99.998% CAS No.: 3396-11-0 Molecular Formula: CsOOCCH3 Formula Weight: 191.95 Physical Form: Lump/Powder Odour: Weak MDL No.: MFCD00013056
Formula: H₃CCOOCs
MW: 191.95 g/mol
Melting Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00013056
CAS Number: 3396-11-0
EINECS: 222-248-0