Potassium acetate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid potassium salt
M-Clarity™ quality level = MQ200
255785-100G 255785-25G 255785-5G
SIAL255785-100GEA 674 EUR
SIAL255785-100G SIAL255785-25G SIAL255785-5G
Potassium acetate, Sigma-Aldrich®
Potassium acetate
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM