Sodium acetate, anhydrous 99%
Supplier: Thermo Scientific
Warning
Synonyms: Acetic acid sodium salt
A13184.0B A13184.0I A13184.30
A13184.0BEA 47.1 EUR
A13184.0B A13184.0I A13184.30
Sodium acetate, anhydrous 99%
Sodium acetate
Purity: 99% CAS No.: 127-09-3 Molecular Formula: C2H3NaO2 Formula Weight: 82.03 pH: 7,5-9 Melting Point: 324° Beilstein Reference: 3595639 Merck Reference: 14,8571 MDL No.: MFCD00012459
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 14,8571