Potassium formate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Formic acid potassium salt
MQ200
294454-1KG 294454-25G 294454-500G
SIAL294454-1KGEA 102 EUR
SIAL294454-1KG SIAL294454-25G SIAL294454-500G
Potassium formate, Sigma-Aldrich®
Potassium formate
Formula: CHKO₂
MW: 84.12 g/mol
Melting Pt: 165…168 °C
Density: 1.99 g/cm³ (20 °C)
MDL Number: MFCD00013100
CAS Number: 590-29-4
EINECS: 209-677-9
Merck Index: 12,07795