Sodium stearate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Stearate sodium, Stearic acid sodium salt
M-Clarity™ quality level = MQ200
S3381-1G S3381-25G S3381-5G
SIALS3381-1GEA 122 EUR
SIALS3381-1G SIALS3381-25G SIALS3381-5G
Sodium stearate, Sigma-Aldrich®
Sodium stearate
Formula: C₁₈H₃₅NaO₂
MW: 306.46 g/mol
Melting Pt: 288 °C
Density: 1.103 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036404
CAS Number: 822-16-2
EINECS: 212-490-5