Sodium octanoate 98%
Supplier: Thermo Scientific
Warning
Synonyms: Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate
334601000 334605000 334600025
ACRO334601000EA 53.9 EUR
ACRO334601000 ACRO334605000 ACRO334600025
Sodium octanoate 98%
Sodium octanoate
Formula: C₈H₁₅NaO₂
MW: 166.2 g/mol
Melting Pt: >225 °C
Density: Min, 0.9949 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00058511
CAS Number: 1984-06-1
EINECS: 217-850-5
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤5 % (K.F.)
Titration with HClO4 ≥97.5 % (on dry substance)