Sodium octanoate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate
M-Clarity™ quality level = MQ200
C5038-100G
C5038-10G
C5038-1KG
C5038-500G
SIALC5038-100GEA
97.6
EUR
SIALC5038-100G
SIALC5038-10G
SIALC5038-1KG
SIALC5038-500G
Sodium octanoate, Sigma-Aldrich®
Sodium octanoate
Formula:
C₈H₁₅NaO₂ MW: 166.2 g/mol Melting Pt: 225…355 °C Density: >0.9949 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00058511 CAS Number: 1984-06-1 EINECS: 217-850-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...