Sodium propionate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Propionic acid sodium salt, Sodiumpropionate
M-Clarity™ quality level = MQ300
W518905-10KG-K W518905-1KG-K W518905-25KG-K
SIALW518905-10KG-KEA 538 EUR
SIALW518905-10KG-K SIALW518905-1KG-K SIALW518905-25KG-K
Sodium propionate, Sigma-Aldrich®
Sodium propionate
Formula: C₃H₅NaO₂
MW: 96.06 g/mol
Melting Pt: 288.76 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002759
CAS Number: 137-40-6
EINECS: 205-290-4
Merck Index: 12,08816