Calcium stearate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
26411-20KG
26411-1KG
SIAL26411-20KGEA
432
EUR
SIAL26411-20KG
SIAL26411-1KG
Calcium stearate, Sigma-Aldrich®
Calcium stearate
Formula:
C₃₆H₇₀CaO₄ Melting Pt: 147…149 °C Density: 1.065 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00036390 CAS Number: 1592-23-0 EINECS: 216-472-8 Merck Index: 13,01708 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...