Magnesium stearate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Stearic acid Magnesium(II) salt, Magnesium(II) Stearate, Stearic acid magnesium salt
M-Clarity™ quality level = MQ100
415057-1KG 415057-25G
SIAL415057-1KGEA 68.3 EUR
SIAL415057-1KG SIAL415057-25G
Magnesium stearate, Sigma-Aldrich®
Magnesium stearate
Formula: C₃₆H₇₀MgO₄
Melting Pt: 130…140 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036391
CAS Number: 557-04-0
EINECS: 209-150-3
Merck Index: 13,05714