Magnesium stearate

Supplier: USP

1374340.EA 0 EUR
1374340.
Magnesium stearate
Magnesium stearate
Formula: C₃₆H₇₀MgO₄
MW: 591,25 g/mol
Melting Pt: 130…140 °C
Density: 1,03 g/cm³ (20 °C)
MDL Number: MFCD00036391
CAS Number: 557-04-0
Merck Index: 13,05714

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR