Calcium propionate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Calcium dipropionate, Propionic acid hemicalcium salt, Propionic acid calcium salt
21230-1KG-F
21230-250G-F
SIAL21230-1KG-FEA
81.5
EUR
SIAL21230-1KG-F
SIAL21230-250G-F
Calcium propionate, Sigma-Aldrich®
Calcium propionate
Formula:
(CH₃CH₂COO)₂Ca MW: 186.22 g/mol Melting Pt: > 300 °C |
MDL Number:
MFCD00036137 CAS Number: 4075-81-4 EINECS: 223-795-8 Merck Index: 13,01703 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...