Thallium(I) acetate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Acetic acid thallium (I) salt, Thallous acetate
T8266-100G T8266-25G T8266-5G
SIALT8266-100GEA 672 EUR
SIALT8266-100G SIALT8266-25G SIALT8266-5G
Thallium(I) acetate, Sigma-Aldrich®
Thallium (I) acetate
Formula: C₂H₃O₂Tl
MW: 263.43 g/mol
Boiling Pt: 1457 °C (1013 hPa)
Melting Pt: 110 °C
Density: 3.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013045
CAS Number: 563-68-8
EINECS: 209-257-5
UN: 1707
ADR: 6.1,II
Merck Index: 12,09392