Copper(I) acetate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
403342-10G 403342-1G
SIAL403342-10GEA 136 EUR
SIAL403342-10G SIAL403342-1G
Copper(I) acetate, Sigma-Aldrich®
Copper (I) acetate
Formula: C₂H₃CuO₂
Melting Pt: 250 °C
MDL Number: MFCD00058908
CAS Number: 598-54-9
EINECS: 209-938-7
Merck Index: 12,02728