Zinc acetate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
383317-100G 383317-25G
SIAL383317-100GEA 331 EUR
SIAL383317-100G SIAL383317-25G
Zinc acetate, Sigma-Aldrich®
Zinc acetate
Formula: C₄H₆O₄Zn
Boiling Pt: 908 °C (1013 hPa)
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012454
CAS Number: 557-34-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 12,10256