4-Aminobutyric acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
A2129-100G A2129-10G A2129-1KG A2129-25G A2129-500G
SIALA2129-100GEA 63.7 EUR
SIALA2129-100G SIALA2129-10G SIALA2129-1KG SIALA2129-25G SIALA2129-500G
4-Aminobutyric acid, Sigma-Aldrich®
4-Aminobutyric acid
Formula: C₄H₉NO₂
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 202 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008226
CAS Number: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429