4-Aminobutyric acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
GABA, γ-Aminobutyric acid
M-Clarity™ quality level = MQ200
A5835-100G
A5835-10G
A5835-10MG
A5835-25G
SIALA5835-100GEA
288
EUR
SIALA5835-100G
SIALA5835-10G
SIALA5835-10MG
SIALA5835-25G
4-Aminobutyric acid, Sigma-Aldrich®
4-Aminobutyric acid
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Boiling Pt: 248 °C (1013 hPa) Melting Pt: 202 °C Density: 1.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008226 CAS Number: 56-12-2 EINECS: 200-258-6 Merck Index: 13,00429 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...