5-Aminovaleric acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: H-5-Ava-OH, 5-Aminopentanoic acid
M-Clarity™ quality level = MQ100
123188-25G 123188-5G
SIAL123188-25GEA 178 EUR
SIAL123188-25G SIAL123188-5G
5-Aminovaleric acid, Sigma-Aldrich®
5-Aminovaleric acid
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 157…158 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008232
CAS Number: 660-88-8
EINECS: 211-544-5