6-Aminocaproic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid
M-Clarity™ quality level = MQ200
07260-100G 07260-1KG 07260-500G
SIAL07260-100GEA 52.4 EUR
SIAL07260-100G SIAL07260-1KG SIAL07260-500G
6-Aminocaproic acid, Sigma-Aldrich®
6-Aminocaproic acid
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 257
Melting Pt: 127
Flash Pt: 207
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS Number: 60-32-2
EINECS: 200-469-3
Merck Index: 13,00431