Glycine
Supplier: PanReac AppliChem
A1067.5000EA 271 EUR
A1067.5000 A1067.1000
Glycine
Glycine
Formula: NH₂CH₂COOH
MW: 75,07 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1,64 g/cm³ (20 °C)
MDL Number: MFCD00008131
CAS Number: 56-40-6
Merck Index: 13,04504
https://si.vwr.com/stibo/web/std.lang.all/19/10/2301910.jpg