Sarcosine, Sigma-Aldrich®
Supplier: Merck
Synonyms: H-Sar-OH, N-Methylglycine, Methylaminoacetic acid
M-Clarity™ quality level = MQ200
131776-100G 131776-500G
SIAL131776-100GEA 46.6 EUR
SIAL131776-100G SIAL131776-500G
Sarcosine, Sigma-Aldrich®
Sarcosine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 63 °C
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00004279
CAS Number: 107-97-1
EINECS: 203-538-6
Merck Index: 12,08519