3-Mercaptopropionic acid, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
M5801-100G M5801-500G M5801-5G
SIALM5801-100GEA 55 EUR
SIALM5801-100G SIALM5801-500G SIALM5801-5G
3-Mercaptopropionic acid, Sigma-Aldrich®
3-Mercaptopropionic acid
Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 217
Melting Pt: 17…19 °C
Density: 1.2199 g/cm³ (20 °C)
Flash Pt: 93
Storage Temperature: Ambient
MDL Number: MFCD00004897
CAS Number: 107-96-0
EINECS: 203-537-0
UN: 2922
ADR: 8 II