Sebacic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Decanedioic acid, 1,8-Octanedicarboxylic acid, 1,10-Decanedioic acid
M-Clarity™ quality level = MQ200
283258-250G 283258-1KG 283258-5G
SIAL283258-250GEA 74.6 EUR
SIAL283258-250G SIAL283258-1KG SIAL283258-5G
Sebacic acid, Sigma-Aldrich®
Sebacic acid
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 374 °C (1013 hPa)
Melting Pt: 127…131 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
REACH: 01-2119519212-52
Merck Index: 12,08558