1,14-Tetradecanedioic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Tetradecanedioic acid, 1,12-Dodecanedicarboxylic acid
M-Clarity™ quality level = MQ100
D221201-25G
D221201-5G
SIALD221201-25GEA
265
EUR
SIALD221201-25G
SIALD221201-5G
1,14-Tetradecanedioic acid, Sigma-Aldrich®
1,14-Tetradecanedioic acid
Formula:
C₁₄H₂₆O₄ MW: 258.36 g/mol Boiling Pt: 407 °C (1013 hPa) Melting Pt: 126 °C Density: 1.108 g/cm³ (21 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002743 CAS Number: 821-38-5 EINECS: 212-476-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...