Malonic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,3-Propanedioic acid
M-Clarity™ quality level = MQ200
M1296-100G M1296-2.5KG M1296-500G M1296-5G
SIALM1296-100GEA 61.4 EUR
SIALM1296-100G SIALM1296-2.5KG SIALM1296-500G SIALM1296-5G
Malonic acid, Sigma-Aldrich®
Malonic acid
Formula: C₃H₄O₄
MW: 104.06 g/mol
Boiling Pt: 387
Melting Pt: 136
Density: 1.669 g/cm³ (20 °C)
Flash Pt: 172
Storage Temperature: Ambient
MDL Number: MFCD00002707
CAS Number: 141-82-2
EINECS: 205-503-0
Merck Index: 13,05732