Succinic acid
Supplier: Molekula
Warning
Synonyms: 1,4-Butanedioic acid
27450488-5KG 27450488-1KG
MOLE27450488-5KGEA 809 EUR
MOLE27450488-5KG MOLE27450488-1KG
Succinic acid
Succinic acid
Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235
Melting Pt: 182
Density: 1.56 g/cm³ (20 °C)
Flash Pt: 206
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8 III
Merck Index: 13,08953
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM