Succinic acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,4-Butanedioic acid
MQ200
14080-1KG
14080-500G
14080-50G
SIAL14080-1KGEA
97.2
EUR
SIAL14080-1KG
SIAL14080-500G
SIAL14080-50G
Succinic acid, Sigma-Aldrich®
Succinic acid
Formula:
HOOCCH₂CH₂COOH MW: 118.09 g/mol Boiling Pt: 235 Melting Pt: 182 Density: 1.56 g/cm³ (20 °C) Flash Pt: 206 Storage Temperature: Ambient |
MDL Number:
MFCD00002789 CAS Number: 110-15-6 EINECS: 203-740-4 UN: 3261 ADR: 8 III Merck Index: 13,08953 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...