Succinic acid, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 1,4-Butanedioic acid

MQ200

S7501-1KG S7501-500G S7501-5KG
SIALS7501-1KGEA 94.9 EUR
SIALS7501-1KG SIALS7501-500G SIALS7501-5KG
Succinic acid, Sigma-Aldrich®
Succinic acid
Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235
Melting Pt: 182
Density: 1.56 g/cm³ (20 °C)
Flash Pt: 206
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8 III
Merck Index: 13,08953
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR