Glutaric acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1,5-Pentanedioic acid

M-Clarity™ quality level = MQ200

G3407-100G G3407-25G G3407-500G
SIALG3407-100GEA 71.4 EUR
SIALG3407-100G SIALG3407-25G SIALG3407-500G
Glutaric acid, Sigma-Aldrich®
Glutaric acid
Formula: C₅H₈O₄
MW: 132.12 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 98 °C
Density: 1.43 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004410
CAS Number: 110-94-1
EINECS: 203-817-2
Merck Index: 13,04486

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR