Adipic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Hexanedioc acid, 1,6-Hexanedioic acid

M-Clarity™ quality level = MQ100

09582-250G 09582-50G
SIAL09582-250GEA 47.2 EUR
SIAL09582-250G SIAL09582-50G
Adipic acid, Sigma-Aldrich®
Adipic acid
Formula: HOOC(CH₂)₄COOH
MW: 146.14 g/mol
Boiling Pt: 337
Melting Pt: 151
Density: 1.36 g/cm³ (25 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
Merck Index: 13,00163
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ac39e8ebd69527d91cb2d15a469a2a69506785&vid=ef907d9f9fd34903274c42b99d1d1ac1a8db6dce&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR