Adipic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Hexanedioc acid, 1,6-Hexanedioic acid
A26357-1KG A26357-3KG A26357-100G A26357-25G A26357-500G
SIALA26357-1KGEA 43.8 EUR
SIALA26357-1KG SIALA26357-3KG SIALA26357-100G SIALA26357-25G SIALA26357-500G
Adipic acid, Sigma-Aldrich®
Adipic acid
Formula: HOOC(CH₂)₄COOH
MW: 146.14 g/mol
Boiling Pt: 337
Melting Pt: 151
Density: 1.36 g/cm³ (25 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
Merck Index: 13,00163
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ac39e8ebd69527d91cb2d15a469a2a69506785&vid=ef907d9f9fd34903274c42b99d1d1ac1a8db6dce&prid=web&clid=SAPDAM