Pimelic acid 98%
Supplier: Thermo Scientific
Warning
Synonyms: Heptanedioic acid, 1,5-Pentanedicarboxylic acid, 1,7-Heptanedioic acid
131230250 131231000 131235000
ACRO131230250EA 33.1 EUR
ACRO131230250 ACRO131231000 ACRO131235000
Pimelic acid 98%
Pimelic acid
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 212 °C (10 mmHg)
Melting Pt: 103…106 °C
Density: 1,329 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004425
CAS Number: 111-16-0
EINECS: 203-840-8
Merck Index: 12,07585
Order Now
Specification Test Results
Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 103 to 106 °C
Titration with NaOH 97.5 to 102.5 %